Learn More
Perkin Elmer US LLC Burgener T2100 PTFE Nebulizer, 0.5-3.0 mL/min
Catalog No. 502113240
Encompass_Preferred
This PTFE T2100 nebulizer can handle very high level of particles in sample and fits regular chambers as a direct replacement for glass concentric nebulizers. It operates on standard pressures of 30 - 45 psi and can be used for any liquid. Salting is very rare, but can occur over long periods of time, especially with high Sodium levels in the solution. Such salting can be cleared easily with a dip in 5 - 10% HF.
Catalog No. 50-211-3240 Supplier Perkin Elmer US LLC Supplier No. N0777033
This PTFE T2100 nebulizer can handle very high level of particles in sample and fits regular chambers as a direct replacement for glass concentric nebulizers. It operates on standard pressures of 30 - 45 psi and can be used for any liquid. Salting is very rare, but can occur over long periods of time, especially with high Sodium levels in the solution. Such salting can be cleared easily with a dip in 5 - 10% HF.