Learn More
Medchemexpress LLC Burixafor (hydrobromide) | 1191450-19-7 | 98.0% | 769.01 | 1 ML
SDP
Catalog No. 5000401884
Encompass_Preferred
Burixafor hydrobromide is a potent CXCR4 antagonist that inhibits the binding of CXCL12 to CXCR4 and blocks downstream signaling. It mobilizes CD34+ hematopoietic stem/progenitor cells from bone marrow to peripheral blood. This compound is used for research on autologous hematopoietic stem cell transplantation (ASCT).
  • Potent CXCR4 antagonist with a pIC50 of 7.4.
  • Inhibits binding of CXCL12 to CXCR4.
  • Mobilizes CD34+ hematopoietic stem/progenitor cells.
  • Useful for research on autologous hematopoietic stem cell transplantation.
Catalog No. 50-004-01884 Supplier Medchemexpress LLC Supplier No. HY19867A10MM/1ML
Burixafor hydrobromide is a potent CXCR4 antagonist that inhibits the binding of CXCL12 to CXCR4 and blocks downstream signaling. It mobilizes CD34+ hematopoietic stem/progenitor cells from bone marrow to peripheral blood. This compound is used for research on autologous hematopoietic stem cell transplantation (ASCT).
  • Potent CXCR4 antagonist with a pIC50 of 7.4.
  • Inhibits binding of CXCL12 to CXCR4.
  • Mobilizes CD34+ hematopoietic stem/progenitor cells.
  • Useful for research on autologous hematopoietic stem cell transplantation.