Learn More
Medchemexpress LLC Burixafor hydrobromide | 1191450-19-7 | 98.0% | 769.01 | 100 MG
SDP
Catalog No. 5000403308
Encompass_Preferred
Burixafor (TG-0054) hydrobromide is a potent CXCR4 antagonist with a pIC50 of 7.4. It inhibits the binding of CXCL12 to CXCR4, antagonizes CXCL12-induced recruitment of Gαᵢ and β-arrestin2, and blocks the downstream Gαᵢ-mediated inhibitory effect on cAMP signal transduction. Burixafor hydrobromide mobilizes CD34+ hematopoietic stem/progenitor cells (HSPC) from the bone marrow to the peripheral blood and can be used for research on autologous hematopoietic stem cell transplantation (ASCT).
  • Potent CXCR4 antagonist with a pIC50 of 7.4
  • Inhibits CXCL12 binding to CXCR4
  • Antagonizes CXCL12-induced recruitment of Gαᵢ and β-arrestin2
  • Blocks downstream Gαᵢ-mediated inhibitory effect on cAMP signal transduction
  • Mobilizes CD34+ hematopoietic stem/progenitor cells (HSPC) from bone marrow to peripheral blood
  • Can be used for research on autologous hematopoietic stem cell transplantation (ASCT)
Catalog No. 50-004-03308 Supplier Medchemexpress LLC Supplier No. HY19867A100MG
Burixafor (TG-0054) hydrobromide is a potent CXCR4 antagonist with a pIC50 of 7.4. It inhibits the binding of CXCL12 to CXCR4, antagonizes CXCL12-induced recruitment of Gαᵢ and β-arrestin2, and blocks the downstream Gαᵢ-mediated inhibitory effect on cAMP signal transduction. Burixafor hydrobromide mobilizes CD34+ hematopoietic stem/progenitor cells (HSPC) from the bone marrow to the peripheral blood and can be used for research on autologous hematopoietic stem cell transplantation (ASCT).
  • Potent CXCR4 antagonist with a pIC50 of 7.4
  • Inhibits CXCL12 binding to CXCR4
  • Antagonizes CXCL12-induced recruitment of Gαᵢ and β-arrestin2
  • Blocks downstream Gαᵢ-mediated inhibitory effect on cAMP signal transduction
  • Mobilizes CD34+ hematopoietic stem/progenitor cells (HSPC) from bone marrow to peripheral blood
  • Can be used for research on autologous hematopoietic stem cell transplantation (ASCT)