Learn More
Medchemexpress LLC Burixafor (hydrobromide) | 1191450-19-7 | 98.0% | 769.01 | 50 MG
SDP
Catalog No. 5000401488
Encompass_Preferred
Burixafor hydrobromide is a potent CXCR4 antagonist with a pIC50 of 7.4. It inhibits CXCL12 binding to CXCR4, antagonizes Gαᵢ and β-arrestin2 recruitment, and blocks downstream Gαᵢ-mediated effects on cAMP signal transduction. This compound mobilizes CD34+ hematopoietic stem/progenitor cells (HSPC) for research on autologous hematopoietic stem cell transplantation (ASCT).
  • Potent CXCR4 antagonist with a pIC50 of 7.4.
  • Inhibits CXCL12 binding to CXCR4.
  • Antagonizes CXCL12-induced Gαᵢ and β-arrestin2 recruitment.
  • Blocks downstream Gαᵢ-mediated inhibitory effect on cAMP signal transduction.
  • Mobilizes CD34+ hematopoietic stem/progenitor cells (HSPC).
  • Useful for research on autologous hematopoietic stem cell transplantation (ASCT).
Catalog No. 50-004-01488 Supplier Medchemexpress LLC Supplier No. HY19867A50MG
Burixafor hydrobromide is a potent CXCR4 antagonist with a pIC50 of 7.4. It inhibits CXCL12 binding to CXCR4, antagonizes Gαᵢ and β-arrestin2 recruitment, and blocks downstream Gαᵢ-mediated effects on cAMP signal transduction. This compound mobilizes CD34+ hematopoietic stem/progenitor cells (HSPC) for research on autologous hematopoietic stem cell transplantation (ASCT).
  • Potent CXCR4 antagonist with a pIC50 of 7.4.
  • Inhibits CXCL12 binding to CXCR4.
  • Antagonizes CXCL12-induced Gαᵢ and β-arrestin2 recruitment.
  • Blocks downstream Gαᵢ-mediated inhibitory effect on cAMP signal transduction.
  • Mobilizes CD34+ hematopoietic stem/progenitor cells (HSPC).
  • Useful for research on autologous hematopoietic stem cell transplantation (ASCT).