Learn More
Medchemexpress LLC Burixafor hydrobromide | 1191450-19-7 | 98.0% | 769.01 | C27H51N8O3P·5/2 HBr | 10 MG
SDP
Catalog No. 5000296747
Encompass_Preferred
Burixafor hydrobromide is the hydrobromide salt of burixafor, a selective, orally active CXCR4 antagonist supplied as a research reagent for preclinical and in vitro studies of CXCR4-mediated biology, including cancer metastasis, angiogenesis, and hematopoietic stem cell mobilization.
  • Selective CXCR4 antagonist for receptor-targeted studies.
  • High purity (98.0%) suitable for research applications.
  • Water soluble (≈50 mg/mL) with ultrasonic assistance.
  • Solid, light yellow to yellow appearance for easy handling.
  • Recommended storage at -20°C under nitrogen for stability.
  • Available in small research quantity (10 MG).
Catalog No. 50-002-96747 Supplier Medchemexpress LLC Supplier No. HY19867A10MG
Burixafor hydrobromide is the hydrobromide salt of burixafor, a selective, orally active CXCR4 antagonist supplied as a research reagent for preclinical and in vitro studies of CXCR4-mediated biology, including cancer metastasis, angiogenesis, and hematopoietic stem cell mobilization.
  • Selective CXCR4 antagonist for receptor-targeted studies.
  • High purity (98.0%) suitable for research applications.
  • Water soluble (≈50 mg/mL) with ultrasonic assistance.
  • Solid, light yellow to yellow appearance for easy handling.
  • Recommended storage at -20°C under nitrogen for stability.
  • Available in small research quantity (10 MG).