Learn More
Medchemexpress LLC HY-19867A 5mg , Burixafor (hydrobromide) CAS:1191450-19-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19867A5MG
Encompass_Preferred
HY-19867A 5mg Burixafor (hydrobromide) CAS: 1191450-19-7 Burixafor hydrobromide (TG-0054 hydrobromide) is an orally bioavailable and potent antagonist of CXCR4 and a well anti-angiogenic drug that is of potential value in treating choroid neovascularization. Burixafor hydrobromide (TG-0054 hydrobromide) mobilizes mesenchymal stem cells, attenuates inflammation, and preserves cardiac systolic function in a porcine model of myocardial infarction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0240