Learn More
ATCC Burkholderia cenocepacia strain LMG 16656 (ATCC BAA-245™)
Catalog No. 502384832
Encompass
Burkholderia cenocepacia strain LMG 16656 (ATCC BAA-245™) is a whole-genome sequenced type strain isolated from a clinical specimen in the UK. This strain is recommended as a test organism for USP <60> testing of Burkholderia cepacia complex in pharmaceutical QC workflows.
Recommended companion product: MicroQuant™ Burkholderia cenocepacia, low CFU (Fisher Cat. No. NC3796842, ATCC BAA-245-LQ-PACK™).
Catalog No. 50-238-4832 Supplier ATCC Supplier No. BAA245
Burkholderia cenocepacia strain LMG 16656 (ATCC BAA-245™) is a whole-genome sequenced type strain isolated from a clinical specimen in the UK. This strain is recommended as a test organism for USP <60> testing of Burkholderia cepacia complex in pharmaceutical QC workflows.
Recommended companion product: MicroQuant™ Burkholderia cenocepacia, low CFU (Fisher Cat. No. NC3796842, ATCC BAA-245-LQ-PACK™).