Learn More
ATCC Burkholderia cepacia
Supplier:  ATCC 25416
Encompass
Burkholderia cepacia strain UCB 717 was isolated from an onion. This whole-genome sequenced bacterial type strain can be used in infectious disease research, media testing, quality control, and respiratory disease research.
Catalog No. 50-238-1846
 Restricted Item