Learn More
Sigma Aldrich Fine Chemicals Biosciences Burkholderia cepacia Selective Agar
Catalog No. NC2735208
Encompass
Burkholderia cepacia Selective Agar | Composition (agar, 15.0 g/L; crystal violet, 0.002 g/L; lactose, 10.000 g/L; phenol red, 0.080 g/L; sodium chloride, 5.0 g/L; sucrose, 10.0 g/L; tryptone, 10.0 g/L; yeast extract, 1.5 g/L), 500g
Catalog No. NC2735208 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 39573500G
Burkholderia cepacia Selective Agar | Composition (agar, 15.0 g/L; crystal violet, 0.002 g/L; lactose, 10.000 g/L; phenol red, 0.080 g/L; sodium chloride, 5.0 g/L; sucrose, 10.0 g/L; tryptone, 10.0 g/L; yeast extract, 1.5 g/L), 500g