Learn More
ATCC Staphylococcus aureus; subsp. aureus; Strain: NSUHS-221
Catalog No. 502379622
Encompass
Methicillin resistant (MRSA). The depositor states that this strain is PFGE type EMRSA, Spa type t032, and pvl negative.
Catalog No. 50-237-9622 Supplier ATCC Supplier No. BAA2765
Add to Cart
Add to Cart
Methicillin resistant (MRSA). The depositor states that this strain is PFGE type EMRSA, Spa type t032, and pvl negative.