Learn More
ATCC Burkholderia multivorans strain LMG 13010 (ATCC BAA-247™)
Supplier:  ATCC BAA247
Encompass
Burkholderia multivorans strain LMG 13010 (ATCC BAA-247™) is a whole-genome sequenced bacterial type strain isolated from a cystic fibrosis patient in Belgium. This strain is recommended as a test organism for USP <60> testing of Burkholderia cepacia complex in pharmaceutical QC workflows.
Recommended companion product: MicroQuant™ Burkholderia multivorans, low CFU (Fisher Cat. No. NC3796844, ATCC BAA-247-LQ-PACK™).
Catalog No. 50-238-4869