Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Burkholderia multivorans derived from ATCC™ BAA-247™
SDP

Catalog No. 12111120 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-111-120 10 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-111-120 Supplier Microbiologics™ Supplier No. 01270E7
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for media QC and presence/absence testing to enumeration testing, water testing, and disinfectant testing.

  • Use a single strain or combine multiple strains for a mixed population.
  • Each quantitative microorganism pellet delivers a specific range of CFU, and can easily manipulate Epower to deliver virtually any concentration.
  • Includes 10 pellets of a single microorganism with a predetermined quantitative assay

Specifications

ATCC® ATCC BAA-247
Biosafety Level 2
Content And Storage 2°C to 8°C
Format Epower
Manufacturing Origin United States
Product Type Quality Control
For Use With (Application) Antimicrobial Effectiveness/Preservation Efficacy Testing
Packaging Type Plastic Box
Quantity 10 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.