Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Burkholderia multivorans Derived From ATCC™ BAA-247™
SDP

Catalog No. 01257542 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-257-542 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-257-542 Supplier Microbiologics™ Supplier No. 01270A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quantitative microorganism preparation that delivers less than 100 CFU per 0.1 mL of hydrated suspension.

  • No dilutions required
  • Kit contains 5 pellets of a single enumerated strain and 5 vials of hydrating fluid (1.2 mL per vial)
  • Delivers 10 to 100 CFU per inoculum

Specifications

ATCC® ATCC™ BAA-247™
Biosafety Level 2
Certifications/Compliance ISO 17034:2016, A2LA Cert. No. 2655.02
Format Kit
Manufacturing Origin United States
Sterility Non-sterile
Target Organism Class Gram-negative Bacteria
For Use With (Application) Growth Promotion and Media Testing
Includes 5 Vials Single, Quantitated Microorganism (1 Lyophilized Pellet/Vial), 5 Vials Hydrating Fluid (1.2 mL/Vial)
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.