Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Braenderup
Supplier:  ATCC BAA664
Encompass
The DNA from this strain is used by laboratories in the Pulsenet system as the molecular size standard for pulsed-field gel electrophoresis (PFGE) analysis of bacterial pathogens (see notes).
Catalog No. 50-238-0191