Learn More
Electron Microscopy Sciences C-Chip C-Chip Hemacytometer Burker
SDP
Catalog No. 501929513
Encompass_Preferred
C-Chip disposable hemacytometer offers precise volume control and is easy use. Features quartz grade optical plastic, two counting chambers are fitted with clearest grid pattern, and continuous sample containment. Use for blood cell, bacteria, spore, and sperm counting. Precise design leads to highly reproducible results.
Catalog No. 50-192-9513 Supplier Electron Microscopy Sciences Supplier No. 6350806
C-Chip disposable hemacytometer offers precise volume control and is easy use. Features quartz grade optical plastic, two counting chambers are fitted with clearest grid pattern, and continuous sample containment. Use for blood cell, bacteria, spore, and sperm counting. Precise design leads to highly reproducible results.