Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Burn Out Solution, Reagents

Catalog No. 01335827
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-827 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-827 Supplier Reagents Holdings Llc Supplier No. CB00990010F
Add to Cart
Add to Cart

Burn Out Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Burn Out Solution
Color Colorless
CAS 7732-18-5, 67-56-1, 67-56-1
Grade Reagent
Packaging Cubitainer
Specific Gravity 0.9579
pH 4 to 6
Quantity 10 L
Sterility Non-sterile
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.