Learn More
Hach Company Burner, Bunsen
Supplier:  Hach Company 2162700
Encompass
Die-cast zinc alloy, with serrated gas inlet connections. Adjustable, with fine pitch threaded needle valve and flame stabilizer. Overall height: 15.6 cm (6.1"). Specifically for use with cylinder (L.P.) gas.
Catalog No. 50-199-5481