Learn More
Perkin Elmer US LLC Burner Gasket Kit
Supplier:  Perkin Elmer US LLC 00470988
Encompass_Preferred
Features and Benefits: 100% titanium burner heads, an exclusive PerkinElmer feature that provides maximum corrosion resistance when analyzing any type of sample Burner chambers constructed of inert, wettable plastic to allow for proper drainage of excess sample and minimize sample carryover of previous analyses Multivaned flow spoilers remove large sample droplets from the nebulized spray and reduce susceptibility to analytical interference Gaskets designed for use with aqueous and organic samples. Choose from synthetic rubber gaskets for aqueous solutions and Corkprene or KALREZ gaskets for organic solutions PinAAcle uses the same Burner Heads as AAnalyst series. The latest design has a hole on the rim for ease of rotation during operation. The handle is provided with the instrument  
Catalog No. NC9843710