Learn More
GSC Go Science Crazy Burner Tripod
SCI_ED
Catalog No. S24635 Shop All GSC Products
Change view
Click to view available options
Diameter (English):
3 in.
Catalog No. Diameter (English)
S24635 3 in.
1 options
This item is not returnable. View return policy
Catalog No. S24635 Supplier GSC Go Science Crazy Supplier No. 4BT5410
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Great design that provides stability by angeling the legs outward.
These burner tripods consist of a cast iron ring and nickel-plated steel support legs. Legs can be removed for storage. Tripod, 3 in diameter, 6 in height.
Specifications
Material Cast Iron, Nickel-plated Steel
Construction Cast-iron ring and removable nickel-plated steel support legs
Diameter (English) 3 in.
Height (English) 6 in.