Learn More
Med Vet International Miltex Dental Burnisher, 1 Double-Ended, 71-20
Catalog No. 502830745
Encompass_Preferred
Burnishers are used to smooth amalgam after condensing, contour matrix bands before placement and begin the contouring of the amalgam. These can also be used with other restorative materials. 1 Double-ended.
Catalog No. 50-283-0745 Supplier Med Vet International Supplier No. MIL7120EA
Burnishers are used to smooth amalgam after condensing, contour matrix bands before placement and begin the contouring of the amalgam. These can also be used with other restorative materials. 1 Double-ended.