Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC AbilityOne Floor Pad, Tan Burnishing, 3400 Series, 14 in Diameter

Catalog No. 502839508
Encompass

Made of 3M floor pad materials, this 11/16 in thick tan burnishing pad is designed for rotary disk floor machines operating above 1500 rpm. It is suitable for use on resilient tile, terrazzo, marble, and ceramic floors. The pads are washable, reusable, and corrosion-resistant, and are designed to produce a "wet look" gloss in high-traffic areas. Each box contains 5 pads.

  • Designed for rotary disk floor machines operating above 1500 rpm.
  • Suitable for use on resilient tile, terrazzo, marble, and ceramic floors.
  • Washable and reusable.
  • Corrosion-resistant.
  • Produces a "wet look" gloss in high-traffic areas.
  • Thickness: 11/16 in.
  • Color: Tan.
  • Diameter: 14 in.
  • Comes in a box of 5 pads.

Catalog No. 50-283-9508 Supplier MSC Supplier No. 89467526
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Made of 3M floor pad materials, this 11/16 in thick tan burnishing pad is designed for rotary disk floor machines operating above 1500 rpm. It is suitable for use on resilient tile, terrazzo, marble, and ceramic floors. The pads are washable, reusable, and corrosion-resistant, and are designed to produce a "wet look" gloss in high-traffic areas. Each box contains 5 pads.

  • Designed for rotary disk floor machines operating above 1500 rpm.
  • Suitable for use on resilient tile, terrazzo, marble, and ceramic floors.
  • Washable and reusable.
  • Corrosion-resistant.
  • Produces a "wet look" gloss in high-traffic areas.
  • Thickness: 11/16 in.
  • Color: Tan.
  • Diameter: 14 in.
  • Comes in a box of 5 pads.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.