Learn More
MSC AbilityOne SKILCRAFT Floor Pad, Aqua, 19 inch Diameter, 3100 Series
Catalog No. 502839530
Encompass
The SKILCRAFT Aqua Burnishing Floor Pad is engineered for professional floor maintenance, designed to buff very soft finishes or polish soft waxes on wood floors. Constructed from durable materials, this 3100 Series pad delivers long-lasting performance for consistent results.
  • Made from durable, non-woven fabric
  • Designed for 175-600 rpm machines
  • Ideal for buffing and polishing soft finishes on wood floors
  • 19 inch diameter
  • Aqua color
  • Delivers long-lasting performance
Catalog No. 50-283-9530 Supplier MSC Supplier No. 89463616
The SKILCRAFT Aqua Burnishing Floor Pad is engineered for professional floor maintenance, designed to buff very soft finishes or polish soft waxes on wood floors. Constructed from durable materials, this 3100 Series pad delivers long-lasting performance for consistent results.
  • Made from durable, non-woven fabric
  • Designed for 175-600 rpm machines
  • Ideal for buffing and polishing soft finishes on wood floors
  • 19 inch diameter
  • Aqua color
  • Delivers long-lasting performance