Learn More
MSC AbilityOne SKILCRAFT Floor Pad, Natural Blend Burnishing, 3500 Series, 17 in Diameter
Catalog No. 502839372
Encompass
This floor pad is constructed from natural hair and synthetic fiber materials, ideal for rotary disk floor machines operating above 1500 rpm. With an 11/16 in thickness, it is suitable for various floor types including resilient tile, terrazzo, marble, and ceramic, offering reusability and corrosion resistance. Each box contains 5 pads.
  • Restores and intensifies the natural shine of floor finishes.
  • Enhances the overall aesthetic appeal through burnishing.
  • Designed to repair floor finishes, bringing out the original shape of the floor.
  • Built for high-speed friction.
  • Suitable for various floor finishes.
Catalog No. 50-283-9372 Supplier MSC Supplier No. 89471965
This floor pad is constructed from natural hair and synthetic fiber materials, ideal for rotary disk floor machines operating above 1500 rpm. With an 11/16 in thickness, it is suitable for various floor types including resilient tile, terrazzo, marble, and ceramic, offering reusability and corrosion resistance. Each box contains 5 pads.
  • Restores and intensifies the natural shine of floor finishes.
  • Enhances the overall aesthetic appeal through burnishing.
  • Designed to repair floor finishes, bringing out the original shape of the floor.
  • Built for high-speed friction.
  • Suitable for various floor finishes.