Learn More
MSC SKILCRAFT Floor Pad, Tan, 27 in, 3400 Series
Catalog No. 502839586
Encompass
This SKILCRAFT Floor Pad is a Tan Burnishing pad from the 3400 Series, engineered for optimal performance with rotary disk floor machines operating above 1500 rpm. It is designed to achieve a 'wet look' high-gloss finish on various floor types, including resilient tile, terrazzo, marble, and ceramic. Each pad is 27 in in diameter, 11/16 in thick, and made from durable 3M™ floor pad materials, supplied in a pack of 5.
  • Designed for high-speed rotary disk floor machines
  • Creates a high-gloss "wet look" finish
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors
  • Made from durable floor pad materials
  • 27 in diameter
  • 11/16 in thick
  • Pack of 5 pads
Catalog No. 50-283-9586 Supplier MSC Supplier No. 89471932
This SKILCRAFT Floor Pad is a Tan Burnishing pad from the 3400 Series, engineered for optimal performance with rotary disk floor machines operating above 1500 rpm. It is designed to achieve a 'wet look' high-gloss finish on various floor types, including resilient tile, terrazzo, marble, and ceramic. Each pad is 27 in in diameter, 11/16 in thick, and made from durable 3M™ floor pad materials, supplied in a pack of 5.
  • Designed for high-speed rotary disk floor machines
  • Creates a high-gloss "wet look" finish
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors
  • Made from durable floor pad materials
  • 27 in diameter
  • 11/16 in thick
  • Pack of 5 pads