Learn More
MSC AbilityOne Floor Pad, Combo Burnishing, 20 inch, Ivory
Catalog No. 502839589
Encompass
These combo burnishing floor pads are designed for use with rotary disk floor machines up to 2500 rpm. They are suitable for cleaning resilient tile, terrazzo, marble, and ceramic floors. Constructed from a polyester and natural hair blend, these ivory pads are 1 inch thick and can be washed, reused, and are corrosion-resistant.
  • Designed for rotary disk floor machines up to 2500 rpm.
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors.
  • Made from a polyester and natural hair blend.
  • Ivory color.
  • 1 inch thickness.
  • Washable, reusable, and corrosion-resistant.
  • Comes in a box of 5 pads.
Catalog No. 50-283-9589 Supplier MSC Supplier No. 89471940
These combo burnishing floor pads are designed for use with rotary disk floor machines up to 2500 rpm. They are suitable for cleaning resilient tile, terrazzo, marble, and ceramic floors. Constructed from a polyester and natural hair blend, these ivory pads are 1 inch thick and can be washed, reused, and are corrosion-resistant.
  • Designed for rotary disk floor machines up to 2500 rpm.
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors.
  • Made from a polyester and natural hair blend.
  • Ivory color.
  • 1 inch thickness.
  • Washable, reusable, and corrosion-resistant.
  • Comes in a box of 5 pads.