Learn More
MSC ABILITYONE Floor Pad, Aqua, Burnishing, 3100 Series, 17 in Diameter
Catalog No. 502839656
Encompass
This floor pad is designed for aqua burnishing, part of the 3100 Series, and features durable 3M floor pad materials for consistent performance. It is ideal for buffing and polishing delicate finishes on wood floors.
  • Made of 3M floor pad materials that deliver long-lasting performance.
  • Used to buff very soft finishes or polish soft waxes on wood floors.
  • Designed for 175-600 rpm.
  • 5 pads per box.
Catalog No. 50-283-9656 Supplier MSC Supplier No. 89463715
This floor pad is designed for aqua burnishing, part of the 3100 Series, and features durable 3M floor pad materials for consistent performance. It is ideal for buffing and polishing delicate finishes on wood floors.
  • Made of 3M floor pad materials that deliver long-lasting performance.
  • Used to buff very soft finishes or polish soft waxes on wood floors.
  • Designed for 175-600 rpm.
  • 5 pads per box.