Learn More
MSC AbilityOne Floor Pad, Tan Burnishing, 3400 Series, 22 in Diameter
Catalog No. 502839592
Encompass
This tan burnishing floor pad is made of 3M floor pad materials and is 11/16 in thick. It is designed for use with rotary disk floor machines operating above 1500 rpm. The pad can be used on resilient tile, terrazzo, marble, and ceramic floors to produce a "wet look" gloss in high-traffic areas.
  • Pads are washable and reusable.
  • Corrosion-resistant construction.
  • Produces a "wet look" gloss.
  • Suitable for high-traffic areas.
  • Sold in boxes of 5 pads.
Catalog No. 50-283-9592 Supplier MSC Supplier No. 89463665
This tan burnishing floor pad is made of 3M floor pad materials and is 11/16 in thick. It is designed for use with rotary disk floor machines operating above 1500 rpm. The pad can be used on resilient tile, terrazzo, marble, and ceramic floors to produce a "wet look" gloss in high-traffic areas.
  • Pads are washable and reusable.
  • Corrosion-resistant construction.
  • Produces a "wet look" gloss.
  • Suitable for high-traffic areas.
  • Sold in boxes of 5 pads.