Learn More
MSC AbilityOne Floor Pad, Natural Blend Burnishing, 17 inch Diameter, 3500 Series
Catalog No. 502839593
Encompass
This floor pad is designed for high-speed burnishing applications, suitable for various floor types including resilient tile, terrazzo, marble, and ceramic floors. It is made from a blend of natural hair and synthetic fibers, engineered to restore and intensify the natural shine of floor finishes.
  • Designed for rotary disk floor machines operating above 1500 rpm.
  • Made with natural hair and synthetic fiber materials.
  • Enhances overall aesthetic appeal by restoring natural shine.
  • Capable of repairing floor finishes to bring out their original luster.
  • Washable, reusable, and corrosion-resistant.
  • Supplied in a box of 5 pads.
Catalog No. 50-283-9593 Supplier MSC Supplier No. 89464804
This floor pad is designed for high-speed burnishing applications, suitable for various floor types including resilient tile, terrazzo, marble, and ceramic floors. It is made from a blend of natural hair and synthetic fibers, engineered to restore and intensify the natural shine of floor finishes.
  • Designed for rotary disk floor machines operating above 1500 rpm.
  • Made with natural hair and synthetic fiber materials.
  • Enhances overall aesthetic appeal by restoring natural shine.
  • Capable of repairing floor finishes to bring out their original luster.
  • Washable, reusable, and corrosion-resistant.
  • Supplied in a box of 5 pads.