Learn More
MSC SKILCRAFT Floor Pad, Eraser Burnishing Pad, 3600 Series, 27 in Diameter
Catalog No. 502839598
Encompass
This 11/16 in thick eraser burnishing pad is designed for rotary disk floor machines operating above 1500 rpm. It is suitable for use on resilient tile, terrazzo, marble, and ceramic floors. The pads are washable, reusable, and corrosion-resistant, effectively removing scuffs and marks. Each box contains 5 pads.
  • Prepares finished floors for optimal gloss.
  • Provides light scrubbing to remove surface-level dirt, scuff marks, and other imperfections.
  • Formulated for minimal finish removal.
  • Effectively lifts surface-level soil.
  • Excellent choice for daily cleaning.
Catalog No. 50-283-9598 Supplier MSC Supplier No. 89467484
This 11/16 in thick eraser burnishing pad is designed for rotary disk floor machines operating above 1500 rpm. It is suitable for use on resilient tile, terrazzo, marble, and ceramic floors. The pads are washable, reusable, and corrosion-resistant, effectively removing scuffs and marks. Each box contains 5 pads.
  • Prepares finished floors for optimal gloss.
  • Provides light scrubbing to remove surface-level dirt, scuff marks, and other imperfections.
  • Formulated for minimal finish removal.
  • Effectively lifts surface-level soil.
  • Excellent choice for daily cleaning.