Learn More
MSC AbilityOne Floor Pads, 20 inch Diameter, Tan, Buffing
Catalog No. 502839647
Encompass
SKILCRAFT Floor Pads are designed for buffing resilient tile, terrazzo, marble, and ceramic floors. These 20-inch diameter pads are washable, reusable, and corrosion-resistant, designed not to leave any residue, and suitable for use with rotary disk floor machines rated between 150-350 rpm.
  • Washable, reusable, and corrosion-resistant pads
  • Designed not to leave any residue
  • Versatile for both buffing and cleaning tasks
  • Ideal for resilient tile, terrazzo, marble, and ceramic floors
  • For use with rotary disk floor machines rated between 150-350 rpm
Catalog No. 50-283-9647 Supplier MSC Supplier No. 89467542
SKILCRAFT Floor Pads are designed for buffing resilient tile, terrazzo, marble, and ceramic floors. These 20-inch diameter pads are washable, reusable, and corrosion-resistant, designed not to leave any residue, and suitable for use with rotary disk floor machines rated between 150-350 rpm.
  • Washable, reusable, and corrosion-resistant pads
  • Designed not to leave any residue
  • Versatile for both buffing and cleaning tasks
  • Ideal for resilient tile, terrazzo, marble, and ceramic floors
  • For use with rotary disk floor machines rated between 150-350 rpm