Learn More
MSC ABILITYONE Floor Pad, Eraser Burnishing Pad, 3600 Series, 16 in Diameter
Supplier: MSC 89467559
This 11/16 in thick eraser burnishing pad is designed for rotary disk floor machines above 1500 rpm, suitable for resilient tile, terrazzo, marble, and ceramic floors. Pads are washable, reusable, corrosion-resistant, and remove scuffs. Engineered to prepare finished floors for optimal gloss, it provides light scrubbing to eliminate surface-level dirt and imperfections. Its formulation ensures minimal finish removal, making it effective at lifting surface-level soil and an excellent choice for daily cleaning. Each box contains 5 pads.
- Designed to prepare finished floors for optimal gloss.
- Provides light scrubbing to remove surface-level dirt, scuff marks, and other imperfections.
- Formulated for minimal finish removal.
- Effective at lifting surface-level soil.
- Excellent choice for daily cleaning.
- Pads are washable, reusable, and corrosion-resistant.