Learn More
MSC AbilityOne Floor Pad, Eraser Burnishing, 3600 Series, 21 Inch Diameter, 3M Material
Catalog No. 502839725
Encompass
This eraser burnishing floor pad is a 3600 Series pad with a 21 in diameter and 11/16 in thickness. Made with 3M floor pad materials, it is specifically designed for rotary disk floor machines operating above 1500 rpm. It is suitable for use on resilient tile, terrazzo, marble, and ceramic floors, and can also be used to buff very soft finishes or polish soft waxes on wood floors.
  • Washable and reusable
  • Resistant to corrosion
  • Effectively removes scuffs and marks from floors
  • Prepares finished floors for optimal gloss
  • Provides light scrubbing to remove surface-level dirt and imperfections
  • Formulated for minimal finish removal
  • Lifts surface-level soil effectively
Catalog No. 50-283-9725 Supplier MSC Supplier No. 89467450
This eraser burnishing floor pad is a 3600 Series pad with a 21 in diameter and 11/16 in thickness. Made with 3M floor pad materials, it is specifically designed for rotary disk floor machines operating above 1500 rpm. It is suitable for use on resilient tile, terrazzo, marble, and ceramic floors, and can also be used to buff very soft finishes or polish soft waxes on wood floors.
  • Washable and reusable
  • Resistant to corrosion
  • Effectively removes scuffs and marks from floors
  • Prepares finished floors for optimal gloss
  • Provides light scrubbing to remove surface-level dirt and imperfections
  • Formulated for minimal finish removal
  • Lifts surface-level soil effectively