Learn More
MSC AbilityOne Floor Pad, Aqua Burnishing, 3100 Series, 21 in Diameter
Catalog No. 502839749
Encompass
AbilityOne SKILCRAFT Aqua Burnishing Floor Pads are crafted from 3M floor pad materials, ensuring long-lasting performance. These pads are specifically designed for buffing very soft finishes or polishing soft waxes on wood floors, suitable for machines operating at 175 to 600 rpm. Each box contains 5 pads.
  • Used to buff very soft finishes or polish soft waxes on wood floors.
  • Designed for 175 to 600 rpm.
  • Constructed from high-quality floor pad materials.
  • Aqua burnishing pad.
  • 21 in diameter.
  • Comes in a box of 5 pads.
Catalog No. 50-283-9749 Supplier MSC Supplier No. 89464838
AbilityOne SKILCRAFT Aqua Burnishing Floor Pads are crafted from 3M floor pad materials, ensuring long-lasting performance. These pads are specifically designed for buffing very soft finishes or polishing soft waxes on wood floors, suitable for machines operating at 175 to 600 rpm. Each box contains 5 pads.
  • Used to buff very soft finishes or polish soft waxes on wood floors.
  • Designed for 175 to 600 rpm.
  • Constructed from high-quality floor pad materials.
  • Aqua burnishing pad.
  • 21 in diameter.
  • Comes in a box of 5 pads.