Learn More
MSC AbilityOne Floor Pad, Aqua Burnishing, 3100 Series, 20 in Diameter
Catalog No. 502837087
Encompass
This floor pad is constructed from high-quality materials, providing long-lasting performance. These aqua burnishing pads are designed for buffing very soft finishes and polishing soft waxes specifically on wood floors.
  • Long-lasting performance
  • Designed for buffing very soft finishes
  • Polishes soft waxes on wood floors
  • Suitable for machines operating at 175 to 600 rpm
  • Comes in a box of 5 pads
  • 20 inch diameter
Catalog No. 50-283-7087 Supplier MSC Supplier No. 89462568
This floor pad is constructed from high-quality materials, providing long-lasting performance. These aqua burnishing pads are designed for buffing very soft finishes and polishing soft waxes specifically on wood floors.
  • Long-lasting performance
  • Designed for buffing very soft finishes
  • Polishes soft waxes on wood floors
  • Suitable for machines operating at 175 to 600 rpm
  • Comes in a box of 5 pads
  • 20 inch diameter