Learn More
MSC SKILCRAFT Floor Pad, Tan Burnishing, 14 inch, 3400 Series
Catalog No. 502837126
Encompass
This tan burnishing floor pad is made of 3M™ floor pad materials and is 11/16 inch thick. Designed for use with rotary disk floor machines operating above 1500 rpm, it is suitable for resilient tile, terrazzo, marble, and ceramic floors. These pads are washable, reusable, and corrosion-resistant, producing a "wet look" gloss in high traffic areas.
  • Tan color
  • 14 inch diameter with 11/16 inch thickness
  • Designed for rotary disk floor machines operating above 1500 rpm
  • Produces a "wet look" gloss in high traffic areas
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors
  • Washable, reusable, and corrosion-resistant pads
  • Compliant with the U.S. Trade Agreement Act (TAA)
Catalog No. 50-283-7126 Supplier MSC Supplier No. 89464663
This tan burnishing floor pad is made of 3M™ floor pad materials and is 11/16 inch thick. Designed for use with rotary disk floor machines operating above 1500 rpm, it is suitable for resilient tile, terrazzo, marble, and ceramic floors. These pads are washable, reusable, and corrosion-resistant, producing a "wet look" gloss in high traffic areas.
  • Tan color
  • 14 inch diameter with 11/16 inch thickness
  • Designed for rotary disk floor machines operating above 1500 rpm
  • Produces a "wet look" gloss in high traffic areas
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors
  • Washable, reusable, and corrosion-resistant pads
  • Compliant with the U.S. Trade Agreement Act (TAA)