Learn More
MSC New Pig Drum Funnel, Polypropylene, 30 & 55 gal Drums, Burpless
Catalog No. 502839836
Encompass
The New Pig drum funnel features a burpless design for fast, smooth liquid transfer into 30 and 55 gallon drums without splashback. Constructed from thick-wall polypropylene, it offers excellent resistance to UV rays, rust, corrosion, and most chemicals. It includes a ridged interior for a level draining surface and a built-in debris strainer to capture large particles.
  • Burpless design vents during pouring for fast, smooth flow with no dangerous splashback.
  • Thick-wall construction resists UV rays, rust, corrosion and most chemicals.
  • Ridged interior creates a level draining surface for containers and enables faster draining.
  • Built-in debris strainer captures large particles.
Catalog No. 50-283-9836 Supplier MSC Supplier No. 93250124
The New Pig drum funnel features a burpless design for fast, smooth liquid transfer into 30 and 55 gallon drums without splashback. Constructed from thick-wall polypropylene, it offers excellent resistance to UV rays, rust, corrosion, and most chemicals. It includes a ridged interior for a level draining surface and a built-in debris strainer to capture large particles.
  • Burpless design vents during pouring for fast, smooth flow with no dangerous splashback.
  • Thick-wall construction resists UV rays, rust, corrosion and most chemicals.
  • Ridged interior creates a level draining surface for containers and enables faster draining.
  • Built-in debris strainer captures large particles.