Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Burrell Scientific Wrist Action™ Laboratory Shaker Accessory, Finger SUREGRIP™ Clamp

Catalog No. 14400140
Change view
Click to view available options
For Use With (Equipment):
Wrist Action Laboratory Shakers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-400-140 Wrist Action Laboratory Shakers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-400-140 Supplier Burrell Scientific Supplier No. 0757770000

missing translation for 'validMainframeMsg'

For use with Wrist Action Laboratory Shaker

  • Maximum adjustability and security
  • Three-prong vinyl-sleeved red grippers
  • Fully adjustable shaft to accommodate various vessels
  • Handle almost any shape flask, bottle or container up to 250mL
  • Material: Aluminum
  • Color: Silver and red

Specifications

Product Type Clamp
For Use With (Equipment) Wrist Action Laboratory Shakers
No. of Positions 2
Warranty and Service Offering 1 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.