Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Burrell Scientific Wrist Action™ Laboratory Shaker Accessory, Top Platform

Catalog No. 14400143
Change view
Click to view available options
No. of Positions:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Positions
14-400-143 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-400-143 Supplier Burrell Scientific Supplier No. 0757653100
Add to Cart
Add to Cart

For use with Wrist Action Laboratory Shaker excluding Magnum

  • Maximize lab space and unit functionality
  • Ability to hold 8 additional vessels (250ml)
  • Fits all Model 75, 85, and 95 shakers
  • Comes with 10 rubber top platform clamps
  • Additional top platform clamps available
  • Material: Aluminum
  • Color: Silver and black

Specifications

Product Type Top Platform
For Use With (Equipment) Wrist Action Laboratory Shaker excluding Magnum
No. of Positions 8
Warranty and Service Offering 1 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.