Learn More
Cayman Chemical α-Amyrin, Purity: ≥95%, CAS: 638-95-9, 10 mg
SDP
Catalog No. 502988257
Encompass_Preferred
A triterpenoid with diverse biological activities; inhibits HIV-1 reverse transcriptase (IC50 = 3.3 µM); inhibits the proliferation of B16 2F2 melanoma cells and LPS-induced NO production in RAW 264.7 macrophages (IC50s = 50 and 8.98 µM, respectively); reduces TPA-induced MPO activity and ear edema in mice (ID50s = 0.45 and 0.31 mg/ear, respectively)
Catalog No. 50-298-8257 Supplier Cayman Chemical Supplier No. 2983110 MG
A triterpenoid with diverse biological activities; inhibits HIV-1 reverse transcriptase (IC50 = 3.3 µM); inhibits the proliferation of B16 2F2 melanoma cells and LPS-induced NO production in RAW 264.7 macrophages (IC50s = 50 and 8.98 µM, respectively); reduces TPA-induced MPO activity and ear edema in mice (ID50s = 0.45 and 0.31 mg/ear, respectively)