Learn More
Grainger Eaton FAZ Bus Bar, 80 A, Two-Pole, 6 Terminals, 10/Pk.
Catalog No. 502795629
Encompass_Preferred
This phase bus bar is a specialized accessory designed for use with FAZ-NA and FAZ-NA-L UL 489 miniature circuit breakers. It provides reliable electrical connection and distribution, supporting efficient system integration in compatible circuit protection setups.
  • Designed as a two-pole, two-phase configuration
  • Features an 80 A amperage rating
  • Equipped with 6 terminals for system connectivity
  • Certified to UL 489 standards
  • Warning: do not cut
Catalog No. 50-279-5629 Supplier Grainger Supplier No. 19YG94
This phase bus bar is a specialized accessory designed for use with FAZ-NA and FAZ-NA-L UL 489 miniature circuit breakers. It provides reliable electrical connection and distribution, supporting efficient system integration in compatible circuit protection setups.
  • Designed as a two-pole, two-phase configuration
  • Features an 80 A amperage rating
  • Equipped with 6 terminals for system connectivity
  • Certified to UL 489 standards
  • Warning: do not cut