Learn More
Grainger Eaton FAZ Accessory Bus Bar, Three-Pole, 80 A, 57 Terminals
Catalog No. 502795883
Encompass_Preferred
The Eaton FAZ accessory bus bar is designed for use with miniature circuit breakers, providing reliable electrical distribution for branch circuit protection applications. This three-pole bus bar features 57 terminals and is rated for 80 A, ensuring efficient power connectivity.
  • 80 A amperage rating
  • Three-pole configuration
  • 57 terminals
  • UL 1077 and UL 489 certified
  • Suitable for reverse feed
Catalog No. 50-279-5883 Supplier Grainger Supplier No. 19YF80
The Eaton FAZ accessory bus bar is designed for use with miniature circuit breakers, providing reliable electrical distribution for branch circuit protection applications. This three-pole bus bar features 57 terminals and is rated for 80 A, ensuring efficient power connectivity.
  • 80 A amperage rating
  • Three-pole configuration
  • 57 terminals
  • UL 1077 and UL 489 certified
  • Suitable for reverse feed