Learn More
CPC Scientific Pyr-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-NHNH2
(trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific LHRH015A
Encompass
Buserelin is a longer acting synthetic peptide analog of the naturally occurring gonadotropin-releasing hormone (GnRH/LHRH). Substitution of glycine in position 6 by D-serine and that of glycinamide in position 10 by ethylamide, leads to a nonapeptide with a significantly enhanced LHRH effect. The effects of buserelin on follicle-stimulating hormone (FSH) and luteinizing hormone (LH) release are approximately 20 to 170 times higher than those of natural LHRH.ONE-LETTER SEQUENCE: Pyr-HWSYs(tBu)-LRP-NHNH2MOLECULAR FORMULA: C58H83N17O13MOLECULAR WEIGHT:1226.39STORAGE CONDITIONS: -20 5CRESEARCH AREA: HormonalREFERENCES: F.Roila, Biomed. Pharmacother., 43, 279 (1989); R.N.Brogden et al., Drugs, 39, 399 (1990); A.Imai et al., Am. J. Obstet. Gynecol., 182, 1142 (2000); Y.Wang et al., Eur. J. Endocrinol., 146, 447 (2002); A.Imai et al., Int. J. Oncol., 20, 987 (2002); J.Franklin et al., J. Endocrinol., 176, 275 (2003); Merck Index, 14th ed., No. 1503, (2006)
Catalog No. 50-210-9665