Learn More
Chemglass Life Sciences Bushing, #11, Internal, Front Seal, Solid, PTFE, #012 Viton O-Ring
SDP
Catalog No. 501531639
Encompass_Preferred
Bushing, #11, Internal, Front Seal, Solid, PTFE, #012 Viton O-Ring.front seal PTFE bushing for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings listed under CG-362.
Catalog No. 50-153-1639 Supplier Chemglass Life Sciences Supplier No. CG36611
Bushing, #11, Internal, Front Seal, Solid, PTFE, #012 Viton O-Ring.front seal PTFE bushing for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings listed under CG-362.