Learn More
Chemglass Life Sciences Chemglass CG-366-10 Bushing, #7 Internal Front Seal, PTFE with #008 O-ring - for internal-thread fittings
SDP
Catalog No. 502733528
Encompass_Preferred
Solid PTFE front-seal internal bushing (CG-366-10) supplied with a #008 fluoroelastomer (FKM) o-ring. This bushing provides a PTFE sealing surface for internally threaded laboratory fittings and is designed for use with internal-thread assemblies.
  • Solid PTFE construction for broad chemical resistance and low friction.
  • Supplied with a #008 fluoroelastomer (FKM) o-ring for reliable sealing.
  • Sized for #7 internal joints and compatible with internal-thread fittings.
  • Intended for use as a front seal in internally threaded sealing assemblies.
  • Available individually for replacement and maintenance.
Catalog No. 50-273-3528 Supplier Chemglass Life Sciences Supplier No. CG36610
Solid PTFE front-seal internal bushing (CG-366-10) supplied with a #008 fluoroelastomer (FKM) o-ring. This bushing provides a PTFE sealing surface for internally threaded laboratory fittings and is designed for use with internal-thread assemblies.
  • Solid PTFE construction for broad chemical resistance and low friction.
  • Supplied with a #008 fluoroelastomer (FKM) o-ring for reliable sealing.
  • Sized for #7 internal joints and compatible with internal-thread fittings.
  • Intended for use as a front seal in internally threaded sealing assemblies.
  • Available individually for replacement and maintenance.