Learn More
Ace Glass, Inc. Bushing adapter, 14/20 female within 24/40 male
SDP
Catalog No. NC9365071
Encompass_Preferred
Bushing adapter for 14/20 female joint within a 24/40 male joint.
Catalog No. NC9365071 Supplier Ace Glass, Inc. Supplier No. 502114
Bushing adapter for 14/20 female joint within a 24/40 male joint.