Learn More
Chemglass Life Sciences #25 Internal Bushing, Solid, PTFE, #220 Viton O-Ring
SDP
Catalog No. 50873532
Encompass_Preferred
#25 Internal Bushing, Solid, PTFE, #220 Viton O-Ring. Solid bushing for use with CG-360 internal glass threads. Supplied with one Viton o-ring.
Catalog No. 50-873-532 Supplier Chemglass Life Sciences Supplier No. CG36305
#25 Internal Bushing, Solid, PTFE, #220 Viton O-Ring. Solid bushing for use with CG-360 internal glass threads. Supplied with one Viton o-ring.