Learn More
Chemglass Life Sciences Bushing, #25, Internal, Front Seal, Solid, PTFE, #212 Viton O-Ring
SDP
Catalog No. 50873546
Encompass_Preferred
Bushing, #25, Internal, Front Seal, Solid, PTFE, #212 Viton O-Ring. New front seal PTFE bushing for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings listed under CG-362.
Catalog No. 50-873-546 Supplier Chemglass Life Sciences Supplier No. CG36613
Bushing, #25, Internal, Front Seal, Solid, PTFE, #212 Viton O-Ring. New front seal PTFE bushing for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings listed under CG-362.