Learn More
Wilmad Labglass BUSHING PTFE FOR 10MM STIR BEA
Catalog No. 50807919
Encompass
Stirrer Bearing, Precision, PTFE, Bushing Only. Replacement PTFE Bushing for LG-9444 series Stirrer Bearings. For use with 10mm OD Stirrer shafts.
Catalog No. 50-807-919 Supplier Wilmad Labglass Supplier No. LG9444144
Stirrer Bearing, Precision, PTFE, Bushing Only. Replacement PTFE Bushing for LG-9444 series Stirrer Bearings. For use with 10mm OD Stirrer shafts.