Learn More
Enterprise Technology Solutions Business Source Catalog Display Rack,Deluxe,Plastic,20-Sheet Capacity,Black
SDP
Catalog No. 5000832983
Encompass_Preferred
Catalog rack includes index tabs and panels that each hold two letter-size documents for a total, 20-sheet capacity. Stable plastic base allows the panels to rotate 360 degrees. Base has rubber feet to help stabilize the rack from sliding on your worksurface. Injection-molded frame is made from polypropylene.
Catalog No. 50-008-32983 Supplier Enterprise Technology Solutions Supplier No. BSN62908
Catalog rack includes index tabs and panels that each hold two letter-size documents for a total, 20-sheet capacity. Stable plastic base allows the panels to rotate 360 degrees. Base has rubber feet to help stabilize the rack from sliding on your worksurface. Injection-molded frame is made from polypropylene.