Learn More
Enterprise Technology Solutions Business Source Filament Tape,FiberglassReinforced,180lb Tensile,3" Core1"x60Yds,White
SDP
Catalog No. 5000833171
Encompass_Preferred
Filament Tape delivers superior performance. Film backing reinforced with fiberglass filaments offers a full 180 lb. tensile (bursting) strength. It meets USPS and UPS specifications. Core measures 3".
Catalog No. 50-008-33171 Supplier Enterprise Technology Solutions Supplier No. BSN64025
Filament Tape delivers superior performance. Film backing reinforced with fiberglass filaments offers a full 180 lb. tensile (bursting) strength. It meets USPS and UPS specifications. Core measures 3".